missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Thrombospondin-3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Thrombospondin-3, was expressed in the hosts and tags indicated.
2430.00 SEK - 27205.00 SEK
Specifications
Accession Number | NP_038719.2 |
---|---|
Gene ID (Entrez) | 21827 |
Name | Thrombospondin-3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16025860
|
enQuireBio™
QP6787-ye-10ug |
10 ÎĽg | N/A | ||||||
16065860
|
enQuireBio™
QP6787-ye-50ug |
50 ÎĽg | N/A | ||||||
16015860
|
enQuireBio™
QP6787-ye-100ug |
100 ÎĽg |
9055.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16045860
|
enQuireBio™
QP6787-ye-200ug |
200 ÎĽg | N/A | ||||||
16055860
|
enQuireBio™
QP6787-ye-500ug |
500 ÎĽg |
25885.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16035860
|
enQuireBio™
QP6787-ye-1mg |
1 mg | N/A | ||||||
Specifications
NP_038719.2 | |
Thrombospondin-3 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
21827 | |
Research Use Only | |
Thbs3 | |
Recombinant Protein | |
Yeast | |
DLQVIDLLTV GESRQMVAVA EKIRTALLTA GDIYLLSTFR LPPKQGGVLF GLYSRQDNTR WLEASVVGKI NKVLVRYQRE DGKVHAVNLQ QAGLADGRTH TALLRLRGPS RPSPGLQLYV DCKLGDQHAG LPALAPIPPA EVSGLEIRTG QKAYLRMQGF VESMKIILGG SMARVGALSE CPFQGDDSIH NAVTSALQSI LGEQTKALVT QLTLFNQILV ELRDDIRDQV KEMSLIRNTI MECQVCGFHE QRSHCSPSPC FRGVDCMEVY EYPGYRCGPC PPGLQGNGTH CDDINECAHA DPCFPGSSCI NTMPGFHCEA CPPGYKG | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse Thrombospondin-3 Protein