missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Deslorelin Protein
Recombinant Protein
1615.00 SEK - 10695.00 SEK
Specifications
Name | Deslorelin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15907958
|
enQuireBio™
QP11661-5mg |
5 mg |
1615.00 SEK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15997948
|
enQuireBio™
QP11661-25mg |
25 mg |
3555.00 SEK
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15987948
|
enQuireBio™
QP11661-100mg |
100 mg |
10695.00 SEK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Deslorelin Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 99.0% as determined by RP-HPLC. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The Deslorelin peptide was lyophilized with no additives. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant other Deslorelin Protein