missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Kinesin-like protein KIF1C Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Kinesin-like protein KIF1C, was expressed in the hosts and tags indicated.

2430.00 SEK - 40175.00 SEK

Specifications

Accession Number NP_665884.1
Gene ID (Entrez) 113886
Name Kinesin-like protein KIF1C Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16002020
View Documents
enQuireBio™
QP6263-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16042020
View Documents
enQuireBio™
QP6263-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16092010
View Documents
enQuireBio™
QP6263-ec-100ug
100 ÎĽg
9055.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16022020
View Documents
enQuireBio™
QP6263-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16032020
View Documents
enQuireBio™
QP6263-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16012020
View Documents
enQuireBio™
QP6263-ec-1mg
1 mg
40175.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_665884.1
Kinesin-like protein KIF1C Protein
Not determined.
Partial (see sequence information for more details).
Rat
His-SUMO
Tris-based buffer, 50% glycerol
113886
Research Use Only
Kif1c
Recombinant Protein
E. coli
MAGASVKVAV RVRPFNARET SQDAKCVVSM QGNTTSIINP KQSRMFLKAS FDYSYWSHTS VEDPQFASQQ QVYRDIGEEM LLHAFEGYNV CIFAYGQTGA GKSYTMMGRQ EPGQQGIVPQ LCEDLFSRVN VNQSAQLSYS VEVSYMEIYC ERVRDLLNPK SRGSLRVREH PILGPYVQDL SKLAVTSYAD IADLMDCGNK ARTVAATNMN ETSSRSHAVF TIVFTQRSHD QLTGLDSEKV SKISLVNLAG SERADSSGAR GMRLKEGANI NK
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Rat Kinesin-like protein KIF1C Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.