Learn More
enQuireBio™ Recombinant Rat PTMA Protein
A DNA sequence encoding the Rattus norvegicus (Rat) PTMA, was expressed in the hosts and tags indicated.
2430.00 SEK - 26260.00 SEK
Specifications
Accession Number | NP_068508.1 |
---|---|
Gene ID (Entrez) | 100359583 |
Name | PTMA Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16038171
|
enQuireBio™
QP9354-ye-10ug |
10 ÎĽg | N/A | ||||||
16078171
|
enQuireBio™
QP9354-ye-50ug |
50 ÎĽg | N/A | ||||||
16028171
|
enQuireBio™
QP9354-ye-100ug |
100 ÎĽg |
9055.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16058171
|
enQuireBio™
QP9354-ye-200ug |
200 ÎĽg | N/A | ||||||
16068171
|
enQuireBio™
QP9354-ye-500ug |
500 ÎĽg |
25885.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16048171
|
enQuireBio™
QP9354-ye-1mg |
1 mg | N/A | ||||||
Specifications
NP_068508.1 | |
PTMA Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
100359583 | |
Research Use Only | |
Ptma | |
Recombinant Protein | |
Yeast | |
SDAAVDTSSE ITTKDLKEKK EVVEEAENGR DAPANGNAQN EENGEQEADN EVDEEEEEGG EEEEEEEEGD GEEEDGDEDE EAEAPTGKRV AEDDEDDDVE TKKQKKTDED D | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
enQuireBio™ Recombinant Rat PTMA Protein