missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Sortilin Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Sortilin, was expressed in the hosts and tags indicated.

2430.00 SEK - 40175.00 SEK

Specifications

Accession Number NP_113955.1
Gene ID (Entrez) 83576
Name Sortilin Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16005340
View Documents
enQuireBio™
QP6716-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16045340
View Documents
enQuireBio™
QP6716-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16095330
View Documents
enQuireBio™
QP6716-ec-100ug
100 ÎĽg
9055.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16025340
View Documents
enQuireBio™
QP6716-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16035340
View Documents
enQuireBio™
QP6716-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16015340
View Documents
enQuireBio™
QP6716-ec-1mg
1 mg
40175.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_113955.1
Sortilin Protein
Not determined.
Full Length
Rat
His-SUMO
Tris-based buffer, 50% glycerol
83576
Research Use Only
Sort1
Recombinant Protein
E. coli
CEENDYTTWL AHSTDPGDYK DGCILGYKEQ FLRLRKSSVC QNGRDYVVAK QPSICPCSLE DFLCDFGYFR PENASECVEQ PELKGHELEF CLYGKEEHLT TNGYRKIPGD RCQGGMNPAR EVKDLKKKCT SNFLNPKKQN SKSSS
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Rat Sortilin Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.