missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae A-agglutinin-binding subunit Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) A-agglutinin-binding subunit, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7176-ye-1mg
This item is not returnable.
View return policy
Specifications
NP_011483.3 | |
S. cerevisiae A-agglutinin-binding subunit Protein | |
Research Use Only | |
AGA2 | |
Recombinant Protein | |
Yeast | |
QELTTICEQI PSPTLESTPY SLSTTTILAN GKAMQGVFEY YKSVTFVSNC GSHPSTTSKG SPINTQYVF | |
Greater than 90% as determined by SDS-PAGE. |
852851 | |
1 mg | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant S. cerevisiae A-agglutinin-binding subunit Protein > 1mg
Spot an opportunity for improvement?Share a Content Correction