missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae A-agglutinin-binding subunit Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) A-agglutinin-binding subunit, was expressed in the hosts and tags indicated.
3490.00 SEK - 31245.00 SEK
Specifications
Accession Number | NP_011483.3 |
---|---|
Gene ID (Entrez) | 852851 |
Name | S. cerevisiae A-agglutinin-binding subunit Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16077570
|
enQuireBio™
QP7176-ye-10ug |
10 ÎĽg | N/A | ||||||
16017580
|
enQuireBio™
QP7176-ye-50ug |
50 ÎĽg | N/A | ||||||
16067570
|
enQuireBio™
QP7176-ye-100ug |
100 ÎĽg |
12935.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16097570
|
enQuireBio™
QP7176-ye-200ug |
200 ÎĽg | N/A | ||||||
16007580
|
enQuireBio™
QP7176-ye-500ug |
500 ÎĽg |
31245.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16087570
|
enQuireBio™
QP7176-ye-1mg |
1 mg |
N/A
|
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_011483.3 | |
S. cerevisiae A-agglutinin-binding subunit Protein | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |
852851 | |
Research Use Only | |
AGA2 | |
Recombinant Protein | |
Yeast | |
QELTTICEQI PSPTLESTPY SLSTTTILAN GKAMQGVFEY YKSVTFVSNC GSHPSTTSKG SPINTQYVF | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant S. cerevisiae A-agglutinin-binding subunit Protein