Learn More
enQuireBio™ Recombinant Sheep IL4 / Interleukin-4 Protein
A DNA sequence encoding the Ovis aries (Sheep) IL4 / Interleukin-4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6224-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_001009313.2 | |
IL4 / Interleukin-4 Protein | |
Research Use Only | |
IL4 | |
Recombinant Protein | |
E. coli | |
HKCDITLEEI IKTLNILTSR KNSCMELPVA DVFAAPKNAT EKETFCRAGI ELRRIYRSHM CLNKFLGGLD RNLSSLASKT CSVNEAKTST STLRDLLERL KTIMREKYSK C | |
Greater than 80% as determined by SDS-PAGE. |
443327 | |
500 μg | |
Not determined. | |
Full Length of Mature. Protein | |
Sheep | |
His-B2M | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Product Title
enQuireBio™ Recombinant Sheep IL4 / Interleukin-4 Protein > 500μg
Spot an opportunity for improvement?Share a Content Correction