missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Sheep IL4 / Interleukin-4 Protein

A DNA sequence encoding the Ovis aries (Sheep) IL4 / Interleukin-4, was expressed in the hosts and tags indicated.

3180.00 SEK - 40175.00 SEK

Specifications

Accession Number NP_001009313.2
Gene ID (Entrez) 443327
Name IL4 / Interleukin-4 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16001720
View Documents
enQuireBio™
QP6224-ec-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16041720
View Documents
enQuireBio™
QP6224-ec-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16091710
View Documents
enQuireBio™
QP6224-ec-100ug
100 μg
11590.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16021720
View Documents
enQuireBio™
QP6224-ec-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16031720
View Documents
enQuireBio™
QP6224-ec-500ug
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16011720
View Documents
enQuireBio™
QP6224-ec-1mg
1 mg
40175.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_001009313.2
IL4 / Interleukin-4 Protein
Not determined.
Full Length of Mature. Protein
Sheep
His-B2M
Tris-based buffer, 50% glycerol
443327
Research Use Only
IL4
Recombinant Protein
E. coli
HKCDITLEEI IKTLNILTSR KNSCMELPVA DVFAAPKNAT EKETFCRAGI ELRRIYRSHM CLNKFLGGLD RNLSSLASKT CSVNEAKTST STLRDLLERL KTIMREKYSK C
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Sheep IL4 / Interleukin-4 Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.