Learn More
enQuireBio™ Recombinant Sheep IL4 / Interleukin-4 Protein
A DNA sequence encoding the Ovis aries (Sheep) IL4 / Interleukin-4, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP6224-ec-50ug
 View more versions of this product
Product Code. 16041720
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
NP_001009313.2
IL4 / Interleukin-4 Protein
Research Use Only
IL4
Recombinant Protein
E. coli
HKCDITLEEI IKTLNILTSR KNSCMELPVA DVFAAPKNAT EKETFCRAGI ELRRIYRSHM CLNKFLGGLD RNLSSLASKT CSVNEAKTST STLRDLLERL KTIMREKYSK C
Greater than 80% as determined by SDS-PAGE.
443327
50 μg
Not determined.
Full Length of Mature. Protein
Sheep
His-B2M
Tris-based buffer, 50% glycerol
Certificates
Promotions
Promotions
Product Content Correction
Product Title
enQuireBio™ Recombinant Sheep IL4 / Interleukin-4 Protein > 50μg