Learn More
Trajan Scientific and Medical™ Replacement Plungers for PerkinElmer Series 200 Syringes
Replacement plungers for Series 200 LC autosampler syringes.
Brand:  Trajan Scientific and Medical™ 031833
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15248553
  • 955.00 SEK / Each
Estimated Shipment: 07-06-2024
to see stock.
Description
Description
Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
Specifications
±1%
Replacement Plunger
60 mm
250 μL
Borosilicate Glass (Barrel), PTFE (Plunger)
8 mm
Gas tight
Series 200 LC Autosampler Syringes