Learn More
Trajan Scientific and Medical™ Replacement Plungers for PerkinElmer Series 200 Syringes
Replacement plungers for Series 200 LC autosampler syringes.
955.00 SEK
Specifications
Description | Replacement Plunger |
---|---|
Model | Gas tight |
For Use With (Equipment) | Series 200 LC Autosampler Syringes |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
15248553
|
Trajan Scientific and Medical™
031833 |
250 μL |
955.00 SEK
Each |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10286119
|
Trajan Scientific and Medical™
0318441 |
1 mL |
955.00 SEK
Each |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Designed to match the specific syringe barrel to maintain a gas tight sealSpecifications
Replacement Plunger | |
Gas tight | |
Series 200 LC Autosampler Syringes |