missing translation for 'onlineSavingsMsg'
Learn More

RIP3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17563-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16887474

  • 3665.00 SEK / 150µL
Estimated Shipment: 24-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

RIP3 is a novel member of RIP Ser/Thr kinase family. It is a potent inducer of apoptosis and is an important component of TNFR1 signaling complex. RIP3 consists of an N-terminal RIP-like-Kinase domain and a unique, non-homologous C-terminus responsible for caspase activation and apoptosis induction. RIP3 is expressed in multiple tissues including hematopoietic cells and plays a functional role in the regulation of apoptosis and NF-kB signaling. It is recruited to the TNFR1 signaling complex in a RIP-dependant manner where it induces apoptosis by activating caspases and/or inhibiting TNFR1-induced NF-kB activation. RIP3 negatively regulates the TLR3-Trif-mediated NF-kB signaling pathway by competing with the binding of Trif to RIP1. The human RIP3 gene is localized in the chromosomal region 14q11.2.
Specifications

Specifications

RIP3
Polyclonal
Unconjugated
RIPK3
Receptor interacting protein 3, RIP 3, RIP like protein kinase 3, RIP3, RIPK3
Rabbit
Antigen Affinity Chromatography
RUO
11035, 56532
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9QZL0, Q9Y572
Ripk3
RIP3 Fusion Protein Ag11759
150 μL
Primary
Mouse, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers