missing translation for 'onlineSavingsMsg'
Learn More

RIPK5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20102-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16879584

  • 1355.00 SEK / 20µL
Estimated Shipment: 03-04-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is specific to DSTYK.

This gene encodes a dual serine/threonine and tyrosine protein kinase which is expressed in multiple tissues. It is thought to function as a regulator of cell death. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

RIPK5
Polyclonal
Unconjugated
DSTYK
DSTYK, Dusty PK, Dusty protein kinase, DustyPK, HDCMD38P, KIAA0472, RIP homologous kinase, RIP5, RIPK5, SgK496, Sugen kinase 496
Rabbit
Antigen Affinity Chromatography
RUO
213452, 25778, 304791
-20°C
Liquid
Immunofluorescence, Immunocytochemistry
0.32 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6XUX1, Q6XUX2, Q6XUX3
Dstyk
RIPK5 Fusion Protein Ag13678
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers