Learn More
RNASEH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15606-1-AP-150UL
 View more versions of this product
Product Code. 16824854
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
RNASEH1 belongs to the RNase H family. It contains 1 RNase H domain. RNASEH1 is an endonuclease that specifically degrades the RNA of RNA-DNA hybrids.
Specifications
Specifications
RNASEH1
Polyclonal
Unconjugated
RNASEH1
H1RNA, Ribonuclease H type II, ribonuclease H1, RNase H1, RNASEH1, RNH1
Rabbit
Antigen Affinity Chromatography
RUO
19819, 246243, 298933
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O60930, O70338, Q5BK46
Rnaseh1
RNASEH1 Fusion Protein Ag7990
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RNASEH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated