Learn More
RORC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13205-1-AP-20UL
 View more versions of this product
Product Code. 16881304
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
RORC (RAR related orphan receptor C) is a DNA-binding transcription factor and is a member of the NR1 subfamily of nuclear hormone receptors. The specific functions of RORC are not known; however, studies of a similar gene in mice have shown that this gene may be essential for lymphoid organogenesis and may play an important regulatory role in thymopoiesis. In addition, studies in mice suggest that the protein encoded by the RORC gene may inhibit the expression of Fas ligand and IL2. Diseases associated with RORC include Immunodeficiency 42 and Anus Cancer.
Specifications
Specifications
RORC
Polyclonal
Unconjugated
RORC
NR1F3, Nuclear receptor ROR gamma, Nuclear receptor RZR gamma, RAR related orphan receptor C, RORC, RORG, RZR GAMMA, RZRG, TOR
Rabbit
Antigen Affinity Chromatography
RUO
19885, 368158, 6097
-20°C
Liquid
Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P51449, P51450
Rorc
RORC Fusion Protein Ag3961
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
RORC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated