missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RORC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1395.65 SEK - 3665.00 SEK
Specifications
Antigen | RORC |
---|---|
Concentration | 0.15 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
RORC (RAR related orphan receptor C) is a DNA-binding transcription factor and is a member of the NR1 subfamily of nuclear hormone receptors. The specific functions of RORC are not known; however, studies of a similar gene in mice have shown that this gene may be essential for lymphoid organogenesis and may play an important regulatory role in thymopoiesis. In addition, studies in mice suggest that the protein encoded by the RORC gene may inhibit the expression of Fas ligand and IL2. Diseases associated with RORC include Immunodeficiency 42 and Anus Cancer.Specifications
RORC | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
P51449, P51450 | |
19885, 368158, 6097 | |
RORC Fusion Protein Ag3961 | |
Primary | |
-20°C | |
RORC |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
NR1F3, Nuclear receptor ROR gamma, Nuclear receptor RZR gamma, RAR related orphan receptor C, RORC, RORG, RZR GAMMA, RZRG, TOR | |
Rorc | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RORC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech