missing translation for 'onlineSavingsMsg'
Learn More

Hettich™ Rotor Adapter for Carriers no. 1398 and 1599

Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life

Brand:  Hettich™ 1483A

958.95 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10505740

  • 1040.00 SEK / Each

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

  • Rotor Adapter for Carriers no. 1398 and 1599
  • For use with ROTOFIX 32 A and Universal 320/320 R Centrifuges
Specifications

Specifications

Adapter
4 x 15 mL
120 mm
4
Conical, Blood/Urine
17 mm
Carriers no. 1398 and 1599 with ROTOFIX 32 A and Universal 320/320 R Centrifuges
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hettich™ Rotor Adapter for Carriers no. 1398 and 1599 > 4 x 15mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.