Learn More
RPS6 Mouse anti-Human, Unconjugated, Clone: 1C3E10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66886-1-IG-20UL
 View more versions of this product
Product Code. 16889155
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The RPS6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RPS6. This antibody recognizes human, mouse, rat antigen. The RPS6 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
RPS6
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunofluorescence, Flow Cytometry
1C3E10
Unconjugated
20 μL
6194
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RPS6 Mouse anti-Human, Unconjugated, Clone: 1C3E10, Proteintech > 20 μL