Learn More
RPS6 Mouse anti-Human, Unconjugated, Clone: 1C3E10, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | RPS6 |
---|---|
Clone | 1C3E10 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, Immunofluorescence, Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16889155
|
Proteintech
66886-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16879155
|
Proteintech
66886-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RPS6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RPS6. This antibody recognizes human, mouse, rat antigen. The RPS6 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
RPS6 | |
1000 ÎĽg/mL | |
Monoclonal | |
Mouse | |
6194 | |
Primary |
1C3E10 | |
Western Blot, Immunofluorescence, Flow Cytometry | |
Unconjugated | |
Human, Rat, Mouse | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.