Learn More
RRP8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20129-1-AP-20UL
 View more versions of this product
Product Code. 16879624
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
Essential component of the eNoSC (energy-dependent nucleolar silencing) complex, a complex that mediates silencing of rDNA in response to intracellular energy status and acts by recruiting histone-modifying enzymes. The eNoSC complex is able to sense the energy status of cell: upon glucose starvation, elevation of NAD(+)/NADP(+) ratio activates SIRT1, leading to histone H3 deacetylation followed by dimethylation of H3 at ′Lys-9′ (H3K9me2) by SUV39H1 and the formation of silent chromatin in the rDNA locus. In the complex, RRP8 binds to H3K9me2 and probably acts as a methyltransferase. Its substrates are however unknown.
Specifications
Specifications
RRP8
Polyclonal
Unconjugated
RRP8
Cerebral protein 1, KIAA0409, Nucleomethylin, RRP8
Rabbit
Antigen Affinity Chromatography
RUO
101867, 23378, 308911
-20°C
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence
0.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O43159, Q5U4F0, Q9DB85
Rrp8
RRP8 Fusion Protein Ag13994
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RRP8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated