missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRP8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | RRP8 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16879624
|
Proteintech
20129-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16869624
|
Proteintech
20129-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Essential component of the eNoSC (energy-dependent nucleolar silencing) complex, a complex that mediates silencing of rDNA in response to intracellular energy status and acts by recruiting histone-modifying enzymes. The eNoSC complex is able to sense the energy status of cell: upon glucose starvation, elevation of NAD(+)/NADP(+) ratio activates SIRT1, leading to histone H3 deacetylation followed by dimethylation of H3 at ′Lys-9′ (H3K9me2) by SUV39H1 and the formation of silent chromatin in the rDNA locus. In the complex, RRP8 binds to H3K9me2 and probably acts as a methyltransferase. Its substrates are however unknown.Specifications
RRP8 | |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
O43159, Q5U4F0, Q9DB85 | |
101867, 23378, 308911 | |
RRP8 Fusion Protein Ag13994 | |
Primary | |
-20°C | |
RRP8 |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Cerebral protein 1, KIAA0409, Nucleomethylin, RRP8 | |
Rrp8 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RRP8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech