Learn More
Tocris Bioscience™ (RS)-CPP

Potent NMDA antagonist
2655.00 SEK - 10030.00 SEK
Chemical Identifiers
CAS | 100828-16-8 |
---|---|
Molecular Formula | C8H17N2O5P |
Molecular Weight (g/mol) | 252.207 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18064882
![]() |
Tocris Bioscience™
0173/10 |
10 mg |
2655.00 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30001420
![]() |
Tocris Bioscience™
0173/50 |
50 mg |
10030.00 SEK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent NMDA antagonist.R-enantiomer also available.
Chemical Identifiers
100828-16-8 | |
C8H17N2O5P | |
252.207 |
Specifications
100828-16-8 | |
252.21 | |
252.207 |
C8H17N2O5P | |
>98% | |
(RS)-CPP |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature
Your input is important to us. Please complete this form to provide feedback related to the content on this product.