missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RTP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18973-1-AP-150UL
This item is not returnable.
View return policy
Description
Specifically promotes functional cell surface expression of olfactory receptors, but not of other GPCRs.Specifications
RTP1 | |
Polyclonal | |
Unconjugated | |
RTP1 | |
RTP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
132112, 239766, 288006 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P59025, Q8C8C1 | |
Rtp1 | |
RTP1 Fusion Protein Ag6934 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RTP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction