Learn More
S protein RBD anti-SARS-CoV-2, Clone: AM180, ACROBiosystems
HEK293 Monoclonal Antibody
Brand:  ACROBiosystems SPD-M180-1mg
Encompass_Preferred
 View more versions of this product
Product Code. 17216550
  • 33645.83 SEK / 1mg
This item is not returnable. View return policy
Description
Description
This product is a specific antibody against SARS-CoV-2 Spike protein RBD domain. Cross-reactivity with RBD of other coronaviruses has not been tested.
Specifications
Specifications
S protein RBD
AM180
Lyophilized from 0.22ÎĽm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
1 mg
Primary
SARS-CoV-2
Recombinant Antibodies
IgG1 Îş
ELISA
Unconjugated
Human (HEK293)
RUO
This product is a specific antibody against SARS-CoV-2 Spike protein RBD domain. Cross-reactivity with RBD of other coronaviruses has not been tested.
-20°C
Powder
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
S protein RBD anti-SARS-CoV-2, Clone: AM180, ACROBiosystems