missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SACM1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13033-1-AP-20UL
This item is not returnable.
View return policy
Description
Phosphoinositide phosphatase that hydrolyzes PtdIns(3)P and PtdIns(4)P. Has low activity towards PtdIns(3,5)P2.Specifications
SACM1L | |
Polyclonal | |
Unconjugated | |
SACM1L | |
KIAA0851, SAC1, SACM1L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116482, 22908, 83493 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Immunocytochemistry | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9EP69, Q9ES21, Q9NTJ5 | |
Sacm1l | |
SACM1L Fusion Protein Ag3682 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SACM1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction