missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SACM1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | SACM1L |
---|---|
Concentration | 0.41 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16881044
|
Proteintech
13033-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16871044
|
Proteintech
13033-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phosphoinositide phosphatase that hydrolyzes PtdIns(3)P and PtdIns(4)P. Has low activity towards PtdIns(3,5)P2.Specifications
SACM1L | |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
Q9EP69, Q9ES21, Q9NTJ5 | |
116482, 22908, 83493 | |
SACM1L Fusion Protein Ag3682 | |
Primary | |
-20°C | |
SACM1L |
0.41 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KIAA0851, SAC1, SACM1L | |
Sacm1l | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SACM1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech