Learn More
Sartorius Safe-Cone Filters
Prevents aerosols or fluids from contaminating samples and pipets. Sartorius Safe-Cone Filters are unique and replaceable polyethylene filters that increase the efficiency of pipetting and the integrity of samples at the same time. Available in Standard and Plus for different lab applications.
528.00 SEK - 662.00 SEK
Specifications
Color White
For Use With Pipette Tips
Material Polyethylene
Quantity 50/Pk.
View More Specs
Products 9
Product Code Brand Diameter (Metric) Outer Type For Use With (Application) For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer Type For Use With (Application) For Use With (Equipment) Price Quantity & Availability  
10552673
View Documents Promotion Details
Sartorius
721008
2.51 mm Standard General applications 100μL Pipette
621.00 SEK
Pack of 50
Only null left
In Stock
Edge
Add to basket
10708082
View Documents Promotion Details
Sartorius
721005
6.73 mm Standard General applications 5000μL Pipette
528.00 SEK
Pack of 50
Only null left
In Stock
Edge
Add to basket
10287822
View Documents Promotion Details
Sartorius
721017
3.15 mm Plus Radioactive work, cell culture, bacterial and virological work, molecular biology 200/300μL Pipette
646.00 SEK
Pack of 50
Only null left
In Stock
Add to basket
10512845
View Documents Promotion Details
Sartorius
721007
3.15 mm Standard General applications 100μL Pipette
631.00 SEK
Pack of 50
Only null left
In Stock
Edge
Add to basket
10529642
View Documents Promotion Details
Sartorius
721014
1.83 mm Standard General applications 20/50μL Pipette
621.00 SEK
Pack of 50
Only null left
In Stock
Add to basket
10043461
View Documents Promotion Details
Sartorius
721006
5.33 mm Standard General applications 1000/1200μL Pipette
621.00 SEK
Pack of 50
Only null left
In Stock
Edge
Add to basket
10080482
View Documents Promotion Details
Sartorius
721016
5.33 mm Plus Radioactive work, cell culture, bacterial and virological work, molecular biology 1000μL Pipette
646.00 SEK
Pack of 50
Only null left
In Stock
Add to basket
10004042
View Documents Promotion Details
Sartorius
721018
2.51 mm Plus Radioactive work, cell culture, bacterial and virological work, molecular biology 200/300μL Pipette
646.00 SEK
Pack of 50
Only null left
In Stock
Add to basket
10217782
View Documents Promotion Details
Sartorius
721015
6.73 mm Plus Radioactive work, cell culture, bacterial and virological work, molecular biology 5000μL Pipette
662.00 SEK
Pack of 50
Only null left
In Stock
Add to basket
Description
Description
Protection for pipets and samples
  • Acts as a final barrier to the internal components of the pipet
  • Protects the pipet and the sample from contamination
  • Prolongs the pipet's lifetime and reduces maintenance intervals
  • Cost-effective, replaceable filters constructed of polyethylene
Available in two formats:
  • Standard Safe-Cone Filters: For general applications and can be used for the same type of work as the Plus Filter, but need to be changed more frequently
  • Plus Safe-Cone Filters: Self-sealing filters meant for more demanding applications, such as radioactive work, cell culture, bacterial and virological work, and molecular biology
Change the filter as needed
  • The frequency of changing the filter depends on the application and the sample
  • Should be changed daily (or after 50 to 250 pipettings) or immediately in case of over-aspiration
  • To ensure safety of the user, forceps should be used to change the filter
  • In addition, clean the tip cone with ethanol (70%) prior to assembling a new filter
Specifications
Specifications
White
Pipette Tips
Polyethylene
50/Pk.
Documents
Product Certifications
Product Content Correction
Product Title
Sartorius Safe-Cone Filters