Learn More
SCAT Safety Waste Cap 'LISA', V3.0 Blind Plug
Safety Waste Cap 'LISA', V3.0 Blind Plug is an accessory suitable for safely closing unused threaded holes in devices for the safe disposal of liquid HPLC/UHPLC waste.
333.00 SEK - 685.00 SEK
Specifications
For Use With HPLC / UHPLC Systems
Material PTFE
View More Specs
Products 4
Product Code Brand Thread Size Type Includes Price Quantity & Availability  
Product Code Brand Thread Size Type Includes Price Quantity & Availability  
17316429
View Documents
SCAT
107680
GL14 Electrically Conductive Blind plug
424.00 SEK
Each
Only null left
Estimated Shipment: 01-07-2025
Add to basket
17326429
View Documents
SCAT
160523
NPT 1/8 in. Electrically Conductive Blind plug
333.00 SEK
Each
Only null left
Estimated Shipment: 01-07-2025
Add to basket
17336429
View Documents
SCAT
350110
M30x35 Not Electrically Conductive Satellite Blind Plug
610.00 SEK
Each
Only null left
Estimated Shipment: 04-07-2025
Add to basket
17366429
View Documents
SCAT
450110
M30x35 Electrically Conductive Satellite Blind Plug
685.00 SEK
Each
Only null left
Estimated Shipment: 04-07-2025
Add to basket
Description
Description
  • Accessory for Safety Waste Cap 'LISA,' V3.0
  • For extending and attaching various components such as a funnel
  • Made of PTFE for optimal chemical resistance
Specifications
Specifications
HPLC / UHPLC Systems
PTFE
Documents
Product Certifications
Product Content Correction
Product Title
SCAT Safety Waste Cap 'LISA', V3.0 Blind Plug