Learn More
Invitrogen™ SARS-COV-2 3CL Protease (aa1-306), His and Avi Tag Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP87699
Description
Animal Origin Free Storage: This product is stable at -70° for up to 1 year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store at recommended temperature.Avoid repeated freeze-thaw cycles.
Severe Acute Respiratory Syndrome (SARS), an emerging disease characterized by atypical pneumonia, has been attributed to a novel coronavirus (SARS-CoV). The SARS 3C-like protease (SARS 3CL (pro)) is a cysteine protease engaging in the proteolytic cleavage of the viral precursor polyprotein to a series of functional proteins required for corona virus replication and is considered an attractive target for therapeutics against SARS.
Specifications
0.579 mg/mL | |
20 mM HEPES with 10% glycerol, 120 mM NaCl, 2 mM TCEP and no preservative; pH 7.4 | |
SARS-COV-2 3 CL Protease (aa1-306), His and Avi Tag | |
E. coli | |
<1.0 EU/ÎĽg | |
SARS 3 CL Protease | |
Recombinant | |
SARS-CoV-2 3 CL Protease, amino acids Ser1-Gln306 with an initial Met and an N-terminal His-tag and Avi-tag | |
E. coli | |
>95% by SDS-PAGE |
Control, Western Blot | |
36.9 kDa | |
1 mg | |
RUO | |
Coronavirus; COVID; COVID 19; COVID-19; SARS Coronavirus; SARS CoV; SARS CoV2; SARS-CoV-2 | |
Unconjugated | |
His-tag, Avi-tag | |
-80°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.