Learn More
SARS-CoV-2 Spike Human anti-Virus, Unconjugated, Clone: AM006415, Proteintech
Human Recombinant Antibody
Brand:  Proteintech 91351-PTG-20UG
 View more versions of this product
Product Code. 16890446
  • 2258.36 SEK / 20µg
Description
The SARS-CoV-2 Spike antibody from Proteintech is a human recombinant antibody generated with virus of virus SARS-CoV-2 Spike. This antibody recognizes virus antigen. The SARS-CoV-2 Spike antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
SARS-CoV-2 Spike
Monoclonal
Unconjugated
20 ÎĽg
43740568
IgG1
Western Blot, ELISA
AM006415
Human
Primary
Virus
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SARS-CoV-2 Spike Human anti-Virus, Unconjugated, Clone: AM006415, Proteintech > 20 ÎĽg