missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 Spike RBD Antibody, R&D Systems™

Mouse Monoclonal Antibody

1910.00 SEK - 4490.00 SEK

Specifications

Antigen Spike RBD
Clone 1035240
Dilution ELISA, Blockade of Receptor-ligand Interaction 50 ug/mL
Applications ELISA, Blocking Assay
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18182201
View Documents
R&D Systems
MAB105406-100
100 ÎĽg
4490.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
18151871
View Documents
R&D Systems
MAB105406-SP
25 ÎĽg
1910.00 SEK
25µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Spike RBD Monoclonal antibody specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for ELISA, Blocking Assay
Specifications

Specifications

Spike RBD
ELISA, Blockade of Receptor-ligand Interaction 50 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 ÎĽm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 ÎĽm filtered solution in PBS.
HEK293 Human embryonic kidney cell -derived SARS-CoV-2 Spike S1, Met1-Tyr674, Accession # YP_009724390.1
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
1035240
ELISA, Blocking Assay
Unconjugated
Mouse
SARS-CoV-2
YP_009724390.1
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SARS-CoV-2 Spike RBD Antibody, R&D Systems™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.