Learn More
Sartorius Qualitative Grade 1291 Filter Papers
These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration.
Brand: Sartorius FT-3-209-110
This item is not returnable.
View return policy
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifications
Disc | |
Cotton Linters, Refined Pulp | |
2 to 3 ÎĽm | |
1291 | |
0.16 mm | |
110 mm |
Conical Funnel | |
2 to 3 ÎĽm | |
180 sec. | |
100/Pk. | |
Qualitative Filter Paper |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Sartorius Qualitative Grade 1291 Filter Papers > Diameter: 110mm
Spot an opportunity for improvement?Share a Content Correction