Learn More
Sca1 (Ly-6A/E) Rat, anti-Mouse, PE, Clone: E13-161.7, STEMCELL Technologies
Rat Monoclonal Antibody
Brand: Stemcell Technologies 60032PE
This item is not returnable.
View return policy
Description
- The E13-161.7 antibody reacts with Sca1 (Stem Cell Antigen-1 or Ly-6A/E), an 18 kDa GPI-linked protein belonging to the lymphocyte activation protein-6 (Ly-6) family
- Sca1 is expressed on the surface of hematopoietic stem and progenitor cells, myeloid cells, and peripheral B and T lymphocytes
- Sca1 expression levels are strongly upregulated in all strains upon cellular activation
- Sca1 is involved in the regulation of T and B cell responses and is believed to play roles in the differentiation, proliferation, and survival of a variety of stem cells
- Sca1 has emerged as a phenotypic marker of choice for identifying and isolating hematopoietic stem and progenitor cells
Specifications
Sca1 (Ly-6A/E) | |
Monoclonal | |
PE | |
200 ÎĽg | |
Mouse | |
Hematopoietic Stem and Progenitor Cells, Mammary Cells |
Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay | |
E13-161.7 | |
Rat | |
Primary | |
IgG2a Îş |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Sca1 (Ly-6A/E) Rat, anti-Mouse, PE, Clone: E13-161.7, STEMCELL Technologies
Spot an opportunity for improvement?Share a Content Correction