Learn More
SEC24D Mouse anti-Human, Unconjugated, Clone: 3G5G9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67409-1-IG-150UL
 View more versions of this product
Product Code. 16890016
  • 4652.90 SEK / 150µL
Description
The SEC24D antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SEC24D. This antibody recognizes human antigen. The SEC24D antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
SEC24D
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunofluorescence, ELISA
3G5G9
Unconjugated
150 μL
9871
IgG2a
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SEC24D Mouse anti-Human, Unconjugated, Clone: 3G5G9, Proteintech > 150 μL