Learn More
SEC24D Mouse anti-Human, Unconjugated, Clone: 3G5G9, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | SEC24D |
---|---|
Clone | 3G5G9 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16800026
|
Proteintech
67409-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16890016
|
Proteintech
67409-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SEC24D antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SEC24D. This antibody recognizes human antigen. The SEC24D antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
SEC24D | |
1000 ÎĽg/mL | |
Monoclonal | |
Mouse | |
9871 | |
Primary |
3G5G9 | |
Western Blot, Immunofluorescence, ELISA | |
Unconjugated | |
Human | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.