Learn More
SEC24D Mouse anti-Human, Unconjugated, Clone: 3G5G9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67409-1-IG-150UL
Description
The SEC24D antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SEC24D. This antibody recognizes human antigen. The SEC24D antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
SEC24D | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunofluorescence, ELISA | |
3G5G9 | |
Unconjugated | |
150 μL | |
9871 | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.