Learn More
SEC63 Mouse anti-Human, Unconjugated, Clone: 1C10E1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67352-1-IG-20UL
 View more versions of this product
Product Code. 16889935
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The SEC63 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SEC63. This antibody recognizes human antigen. The SEC63 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
SEC63
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunofluorescence, ELISA
1C10E1
Unconjugated
20 μL
11231
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SEC63 Mouse anti-Human, Unconjugated, Clone: 1C10E1, Proteintech > 20 μL