missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC63 Mouse anti-Human, Unconjugated, Clone: 1C10E1, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | SEC63 |
---|---|
Clone | 1C10E1 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16889935
|
Proteintech
67352-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16879935
|
Proteintech
67352-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SEC63 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SEC63. This antibody recognizes human antigen. The SEC63 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
SEC63 | |
1000 ÎĽg/mL | |
Monoclonal | |
Mouse | |
11231 | |
Primary |
1C10E1 | |
Western Blot, Immunofluorescence, ELISA | |
Unconjugated | |
Human | |
IgG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SEC63 Mouse anti-Human, Unconjugated, Clone: 1C10E1, Proteintech