Learn More
SEC63 Mouse anti-Human, Unconjugated, Clone: 1C10E1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67352-1-IG-150UL
 View more versions of this product
Product Code. 16879935
  • 4652.90 SEK / 150µL
Description
The SEC63 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SEC63. This antibody recognizes human antigen. The SEC63 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
SEC63
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunofluorescence, ELISA
1C10E1
Unconjugated
150 μL
11231
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SEC63 Mouse anti-Human, Unconjugated, Clone: 1C10E1, Proteintech > 150 μL