missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sec8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11913-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the Asp-Glu-Ala-Asp (DEAD) box protein family. Members of this protein family are putative RNA helicases, and are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. Two transcript variants encoding the same protein have been identified for this gene.Specifications
Sec8 | |
Polyclonal | |
Unconjugated | |
EXOC4 | |
EXOC4, exocyst complex component 4, Exocyst complex component Sec8, KIAA1699, REC8, Sec8, SEC8L1, Sec8p | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116654, 20336, 60412 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35382, Q62824, Q96A65 | |
EXOC4 | |
Sec8 Fusion Protein Ag2514 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sec8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction