missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINA10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11851-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the serpin family. It is predomitly expressed in the liver and secreted in plasma. It inhibits the activity of coagulation factors Xa and XIa in the presence of protein Z, calcium and phospholipid. Mutations in this gene are associated with venous thrombosis. Alternatively spliced transcript variants have been found for this gene.Specifications
SERPINA10 | |
Polyclonal | |
Unconjugated | |
SERPINA10 | |
PZI, Serpin A10, SERPINA10, ZPI | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171154, 217847, 51156 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q62975, Q8R121, Q9UK55 | |
Serpina10 | |
SERPINA10 Fusion Protein Ag2420 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SERPINA10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction