Learn More
Trajan™ Glass Lined Bulkhead Unions with Locknut
Glass lined stainless steel construcion. Bulkhead unions with locknut
Brand: Trajan™ 1236300
This item is not returnable.
View return policy
Description
- Allows plate mounting of union
- For use with fused silica
- Low dead volume
- High temperature applications
Specifications
Bulkhead Union | |
0.35 mm restriction (at one end) | |
Glass-lined Stainless Steel | |
0.5 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Trajan™ Glass Lined Bulkhead Unions with Locknut > Includes: 0.35mm restriction at one end; ID: 0.5mm; Material: glass lined stainless steel
Spot an opportunity for improvement?Share a Content Correction