Learn More
Trajan™ SilTite™ FingerTite Ferrule System Adapters
For gas chromatography systems
Brand: Trajan™ 0736104
This item is not returnable.
View return policy
Description
- Next generation ferrule system for gas chromatography
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifications
Adapter | |
Two Base Seals | |
Agilent Injector Adaptor | |
Agilent GCs, Thermo 1300 Injector |
Product Content Correction
Product Title
Trajan™ SilTite™ FingerTite Ferrule System Adapters > Agilent injector adapter
Spot an opportunity for improvement?Share a Content Correction