Learn More
BRAND™ SilberBrand™ Pellet Pattern Automatic Burette
Without intermediate stopcock; with needle-valve stopcock with PTFE spindle
Brand: BRAND™ 21918
This item is not returnable.
View return policy
Description
Specifications
50 mL | |
0.08 mL | |
±0.08 mL | |
Class B |
Borosilicate Glass | |
Schellbach Stripe | |
Automatic Buret | |
DIN EN ISO 385 |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
BRAND™ SilberBrand™ Pellet Pattern Automatic Burette > Capacity: 50 mL
Spot an opportunity for improvement?Share a Content Correction