Learn More
Silicon Standard Metal Solution 1000ppm, for AAS, Solution Aqueous, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
Silicon Standard Metal Solution 1000ppm, for AAS, Solution Aqueous
Specifications
Specifications
Packaging | HDPE plastic bottle |
Physical Form | Liquid |
Color | Colorless |
Quantity | 500 mL |
Safety and Handling
Product Identifier
- Silicon solution 1000 ppm
Product Content Correction
Product Title
Silicon Standard Metal Solution 1000ppm, for AAS, Solution Aqueous, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction