Learn More
Silver, brazing flux, white paste, Thermo Scientific™
244.00 SEK - 455.73 SEK
Specifications
Chemical Name or Material Silver, brazing flux
Name Note Active range: 540 to 980°
Color White
MDL Number MFCD01868499
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11314068
View Documents
Thermo Scientific Alfa Aesar
041355.TB
113 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11324068
View Documents
Thermo Scientific Alfa Aesar
041355.TD
454 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Silver, brazing flux
Active range: 540 to 980°
White
MFCD01868499
Videos
Safety and Handling
Safety and Handling
GHS H Statement
H360-H315-H319-H335
May damage fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P281-P308+P313-P501c
H360FD
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Silver, brazing flux, white paste, Thermo Scientific™