missing translation for 'onlineSavingsMsg'
Learn More

SIX2 Mouse anti-Canine, Human, Mouse, Porcine, Rat, Clone: 1F9G11, Proteintech

Mouse Monoclonal Antibody

1236.20 SEK - 3313.85 SEK

Specifications

Antigen SIX2
Concentration 1 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Format Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16878135
View Documents Promotion Details
Proteintech
66347-1-IG-150UL
150 μL
3313.85 SEK
150µL
View alternative products
Save Up to 
»
Estimated Shipment: 20-01-2022
Please to purchase this item. Need a web account? Register with us today!
16888135
View Documents Promotion Details
Proteintech
66347-1-IG-20UL
20 μL
1236.20 SEK
20µL
View alternative products
Save Up to 
»
Estimated Shipment: 20-01-2022
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene is a member of the vertebrate gene family which encode proteins homologous to the Drosophila ′sine oculis′ homeobox protein. SIX2 is a transcription factor which, like other members of this gene family, may be involved in limb or eye development.
Specifications

Specifications

SIX2
Immunohistochemistry (Paraffin), Western Blot
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Monoclonal
-20°C
1 mg/mL
Unconjugated
Mouse
Canine, Mouse, Porcine, Rat, Human
IgG2b
Protein A
Antibody
SDS
Product Certifications