Learn More
SLC22A7 Mouse anti-Human, Unconjugated, Clone: 3D3C3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67479-1-IG-20UL
 View more versions of this product
Product Code. 16880126
  • 1645.54 SEK / 20µL
Description
The SLC22A7 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SLC22A7. This antibody recognizes human antigen. The SLC22A7 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
SLC22A7
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, Immunofluorescence
3D3C3
Unconjugated
20 μL
10864
IgG2a
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SLC22A7 Mouse anti-Human, Unconjugated, Clone: 3D3C3, Proteintech > 20 μL