Learn More
SLC25A46 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | SLC25A46 |
---|---|
Concentration | 333 μg/mL |
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16899893
|
Proteintech
12277-1-AP-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16889893
|
Proteintech
12277-1-AP-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SLC25A46 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SLC25A46. This antibody recognizes human, mouse, rat antigen. The SLC25A46 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
SLC25A46 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Human, Rat, Mouse | |
IgG |
333 μg/mL | |
Polyclonal | |
Rabbit | |
91137 | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.